Order Entry
Mexico
ContactUsLinkComponent
Medulla Oblongata Slide
Medulla Oblongata Slide
Catalog # 470182-718
Supplier:  Avantor
undefined
Medulla Oblongata Slide
Catalog # 470182-718
Supplier:  Avantor
Specifications
  • Description:
    Medulla Oblongata
  • Cat. no.:
    470182-718
  • Stain:
    Luxol fast blue and cresyl violet
  • Supplier No.:
    WARD470182-718
  • Orientation:
    Cross section
Specifications
About this item
Located at the Base of the Brainstem
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Demonstrates cellular details and fiber bundles.