Order Entry
Mexico
ContactUsLinkComponent
Mast Cells Slide
Mast Cells Slide
Catalog # 470182-634
Supplier:  Avantor
undefined
Mast Cells Slide
Catalog # 470182-634
Supplier:  Avantor
Specifications
  • Description:
    Mast Cells
  • Cat. no.:
    470182-634
  • Stain:
    Fuchsin and Hematoxylin
  • Supplier No.:
    WARD470182-634
  • Orientation:
    Whole mount
Specifications
About this item
Flat Mount of Mesentery
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Resorcin fuchsin stained to demonstrate mast cells as well as both white and elastic fibers.