Mammary Gland, Inactive, Human Slide
470183-076
: Avantor
- Description:Mammary Gland, Inactive, Human
- Cat. no.:470183-076
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470183-076
- Orientation:Section
Female Reproductive Organ
- Identifying features clearly distinguishable
- Rigorous quality control standards
Human. Abundant connective tissue is visible. The ducts and surrounding loose connective tissue constitute a lobule. The dense connective tissue contains aggregates of adipocytes.