Order Entry
Mexico
Orders LinkContactUsLinkComponent
Ward's® Longitudinal Wave Demonstration
Ward's® Longitudinal Wave Demonstration
Longitudinal Wave Demonstration
Catalog # 470004-950
Supplier:   Avantor
Ward's® Longitudinal Wave Demonstration
Catalog # 470004-950
Supplier:  Avantor
Ward's® Longitudinal Wave Demonstration
Ward's® Longitudinal Wave Demonstration
Longitudinal Wave Demonstration
Price
$2,479.80
Log in for availability
Each (1UN)
Product Details & Documents
This inexpensive model, developed by Tim Ernest, lets you create longitudinal/compression waves by simply pushing a Slinky® along a track.
  • Grades 6 to 9
  • Length: 152.4 cm
It can be used in studies of waves, and sound to demonstrate compression, rarefaction, amplitude, wavelength, echoes, volume and pitch.
Delivery information: Includes instructions.
Specifications
  • Description:
    Longitudinal Wave Demonstration