Order Entry
Mexico
Orders LinkContactUsLinkComponent
Liver Slide
Liver Slide
Liver Slide, Section, Mallory Triple
Catalog # 470182-884
Supplier:   Avantor
Liver Slide
Catalog # 470182-884
Supplier:  Avantor
Liver Slide
Liver Slide
Liver Slide, Section, Mallory Triple
Price
$166.10
Log in for availability
Each (1UN)
Product Details & Documents
Demonstrates pig-specific liver characteristics
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Pig. Demonstrates the lobular structure of the liver through heavily developed connective tissue, a peculiarity of pig liver.
Specifications
  • Description:
    Liver Slide
  • Stain:
    Mallory Triple
  • Orientation:
    Section