Lily, Sporogenous Tissue
Catalog # 470181-246
Supplier: Avantor
Specifications
- Description:Lily, Sporogenous Tissue, cross section
- Cat. no.:470181-246
- Supplier No.:WARD470181-246
Specifications
About this item
Cross section, quadruple stained
- All identifying features are clearly distinguishable
- Rigorous quality control standards
Very young anther. Shows early cellular development prior to meiosis.