Lily Meiosis, Synizesis Slide
Catalog # 470181-248
Supplier: Avantor
Specifications
- Description:Lily Meiosis, Synizesis
- Cat. no.:470181-248
- Stain:Quadruple
- Supplier No.:WARD470181-248
- Orientation:Cross section
Specifications
About this item
Also Called a Synizetic Knot
- Identifying features clearly distinguishable
- Rigorous quality control standards
Chromatin compacts to one side of the nucleus.