Order Entry
Mexico
Orders LinkContactUsLinkComponent
LDPE Dropping Bottles

LDPE Dropping Bottles

LDPE Dropping Bottles, 125 ml
Catalog # 470201-798
Supplier:   BEL-ART PRODUCTS SE
Supplier #:  F11637-0004_SE
LDPE Dropping Bottles
Catalog # 470201-798
LDPE Dropping Bottles
LDPE Dropping Bottles

LDPE Dropping Bottles, 125 ml

Price
$1,080.00
Log in for availability
Paquete de 12 Case de 6 (12UN)
Product Details & Documents

Molded Polyethylene.

  • Screw cap has dropping tip with 0.8 mm opening
  • Snap-on sealer tip
  • Reusable
Specifications
  • Capacity:
    125 ml
  • Description:
    LDPE Dropping Bottles
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.