Lab Scoops
Lab Scoops
Catalog # 470153-820
Supplier: BEL-ART PRODUCTS SE
Supplier #: F36747-0000_SE
Jump to:
Product Details & Documents
Molded polypropylene scoops are rigid, strong, and smooth.
Steam autoclavable at 121°C/250°F. Scoops are 22cc with a 1 1/4" handle.
Delivery information: Package of 12.
Specifications
- Description:Lab Scoops