Kidney Slide
Kidney Slide, Thin section, Hematoxylin and Eosin
Catalog # 470183-696
Supplier: Avantor
Jump to:
Product Details & Documents
Extremely Thin, Plastic Embedded Tissue For Greater Viewing Clarity
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. For detailed study of cortex and medulla. Shows renal corpuscle, tubule, and collecting duct.
Specifications
- Description:Kidney Slide
- Stain:Hematoxylin and Eosin
- Orientation:Thin section