It's All About Volume Kit
It's All About Volume Kit, Teacher's Guide and Student Copymaster, 7
Catalog # 470039-706
Supplier: Avantor
Jump to:
Product Details & Documents
How Much Space Does This Take Up?
- Perfect for Grades 5-8
- 5 Full Activities
Just add water for a fun, hands-on introduction to the measurement of volume. Five activities lead students through volume calculations and the displacement of liquids, using the included fun squirt fish, sponges, clay, beakers, and graduated cylinders. Developed by Jan Pechenik, the kit activities are aligned to national standards.
Includes:
Steel balls
Beakers
Modeling Clay
Nylon Balls
Cylinders
Plastic Fish
Funnels
Cubic Centimeter Cubes
Droppers
Bags
Instructions
Required but Not Provided: Metric Rulers, Paper Towels, Containers of water, Knife (to cut clay), Calculators (optional), Balance (optional).
Specifications
- Description:It's All About Volume Kit
- No. of Lab Groups:7
- Includes:Teacher's Guide and Student Copymaster