Order Entry
Mexico
ContactUsLinkComponent
Iodamoeba butschlii, Trophozoites and Cysts
Iodamoeba butschlii, Trophozoites and Cysts
  470181-866
 :  Avantor
undefined
Iodamoeba butschlii, Trophozoites and Cysts
  470181-866
 :  Avantor
  • Description:
    Iodamoeba butschlii, Trophozoites and cysts
  • Cat. no.:
    470181-866
  • Stain:
    Hematoxylin
  • Supplier No.:
    WARD470181-866
  • Orientation:
    Smear
Non-pathogenic Amoeba.
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Commensal organism living in the large intestine of humans. It possesses a large, commonly off-center karyosome.