Order Entry
Mexico
Orders LinkContactUsLinkComponent
Ward's® Crystal Form Collection
Ward's® Crystal Form Collection
Introductory Crystal Form Collection, 6
Catalog # 470015-404
Supplier:   Avantor
Ward's® Crystal Form Collection
Catalog # 470015-404
Supplier:  Avantor
Ward's® Crystal Form Collection
Ward's® Crystal Form Collection
Introductory Crystal Form Collection, 6
Price
$985.20
Log in for availability
Each (1UN)
Product Details & Documents
Basic Set of Natural Crystals
  • Hand Selected by WARD's Geologists
  • Teach Students the Basic Crystal Forms
The six main crystal systems are represented in this beginning geology student’s introduction to crystal form. The set is housed in a compartmented collection box, and an identification list is included.
Contents:6 numbered samples, 1" x 1"
Isometric Pyrite
Hexagonal Quartz
Tetragonal Zircon
Orthorhombic Danburite
Monoclinic Orthoclase
Triclinic Microcline
Specifications
  • Description:
    Introductory Crystal Form Collection
  • No. of Samples:
    6