Quincke’s Interference Tubes
Quincke’s Interference Tubes
Catalog # 470106-466
Supplier: GSC INTERNATIONAL SE
Supplier #: 904
Jump to:
Product Details & Documents
To Accompany Your Tuning Fork!
- Comes with instructions and suggestions for use.
- Tuning fork is not included.
This apparatus consists of two Y-tubes about 15 mm in diameter, connected with rubber tubing, with one path being approximately one-half wavelength longer than the other. When a C fork (256 vps) is held near one orifice and the path difference is 65 cm, there will be no sound at the other orifice. When either tube is pinched shut, and the tuning fork is sounded at the intake, sound comes through.
Specifications
- Description:Quincke’s Interference Tubes