Instant Ear Thermometer
Instant Ear Thermometer
Catalog # 470018-918
Supplier: GRAHAM-FIELD INC
Supplier #: 2215
Jump to:
Product Details & Documents
This thermometer features one-touch operation and avoids the hassle of oral thermometers.
- Accurate Readout in One Second
- Fever Alarm
The user-friendly instrument yields accurate results within one second of being placed in the patient’s ear. A fever alarm sounds for temperatures above 100.4°F. The thermometer's display indicates measurements in either Celsius or Fahrenheit, and the Nite-Glo display can be seen in any lighting. The thermometer has a range of 93.2°F to 109.4°F and includes a storage case, lithium battery, and guidebook.
Documents
Specifications
- Description:Instant Ear Thermometer