Imhoff Settling Cones
Imhoff Settling Cone
470211-728
: 1000-0010
Determines Settleable Matter in Water
The leakproof cone is fully graduated from 0–1,000 mL for easy estimating at higher intervals, as well as enhanced with a leakproof closure that facilitates withdrawal of sediment for gravimetric measuring and allows for easy cleaning. It is made of unbreakable polycarbonate for long life. Size: 18"L x 4" dia.
- Description:Imhoff Settling Cone
- Cat. No.:470211-728
- Environmental certification:ISO14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Low Manufacturing Impact



