Order Entry
Mexico
ContactUsLinkComponent
Ice Cream Making Kit: A Heat Transfer Experiment
Ice Cream Making Kit: A Heat Transfer Experiment
Catalog # 470104-476
Supplier:  Avantor
undefined
Ice Cream Making Kit: A Heat Transfer Experiment
Catalog # 470104-476
Supplier:  Avantor
Specifications
  • Description:
    Ice Cream Making Kit
  • No. of Lab Groups:
    5
  • Cat. no.:
    470104-476
  • Supplier No.:
    WARD470104-476
Specifications
About this item
Science Has Never Been Sweeter!
  • Perfect for Grades 4-9
  • Instruction and Lesson Plans Included
Using this kit, students use a simple technique to create edible ice cream and learn about heat transfer, phase change, and insulation along the way. The entire activity, from heat transfer to clean-up, can be performed in a single 45-minute lab period. Kit includes enough materials for five lab stations of six students per group, with 20 ice cream recipes included. Each recipe yields approximately 2-3 cups of ice cream. We recommend using a glue gun to apply the pre-cut insulation to the reusable cans.
Includes:
Water Softener Salt
Metal Cans with Lid (2 sizes)
Foam Insulation
Duct Tape
Instructions
Required but Not Provided: Ice, Glue Gun, Measuring Cup or Beakers, Bowls or Cups, Spoons, Blender (optional).