Order Entry
Mexico
Orders LinkContactUsLinkComponent
Hydraulic Hand Dynamometer, FABRICATION ENTERPRISES INC.
Hydraulic Hand Dynamometer, FABRICATION ENTERPRISES INC.
Hydraulic Hand Dynamometer
Catalog # 470019-094
Supplier #:  12-0240
Hydraulic Hand Dynamometer, FABRICATION ENTERPRISES INC.
Catalog # 470019-094
Supplier:  
Hydraulic Hand Dynamometer, FABRICATION ENTERPRISES INC.
Hydraulic Hand Dynamometer, FABRICATION ENTERPRISES INC.
Hydraulic Hand Dynamometer
Discontinued
Product Details & Documents
Used By Professional Physical Therapists for Reliability and Repeatability
Receive accurate grip strength measurements without feeling the handle move, with the hydraulic and bellows operation of this instrument. A five position adjustable handle accommodates any hand size, while a large, dual-scale gauge features a maximum reading indicator. Range: 0-200 lbs in 2 lb increments; 0-90 kg in 5 kg increments. The all metal construction provides durability. Comes in a protective carrying case with manual. Size: 10"L x 2 1/2" dia.
Specifications
  • Description:
    Hydraulic Hand Dynamometer