Helical Spring Set
Catalog # 470149-836
Supplier: STANLEY SPRING & STAMPING
Specifications
- Description:Replacement Spring for Helical Spring Set
- Cat. no.:470149-836
- Supplier No.:6645601B&L
Specifications
About this item
This set consists of a carefully selected steel spring, a mass of 0.5 kg with a hook on both ends, and a mass of 1 kg with a single hook.
The spring has hooks on both ends and is approximately 20 cm long.