Order Entry
Mexico
Orders LinkContactUsLinkComponent
Gold Foil Electroscope
Gold Foil Electroscope
Gold foil electroscope
Catalog # 470020-658
Supplier #:  ELGL01
Gold Foil Electroscope
Catalog # 470020-658
Gold Foil Electroscope
Gold Foil Electroscope
Gold foil electroscope
Price
$1,040.95
Log in for availability
Each (1UN)
Product Details & Documents
A Highly Sensitive Electroscope
  • Highly sensitive gold foil
  • Detect small static charges
This high sensitive electroscope can detect charges that a regular electroscope cannot. A gold leaf is mounted on an insulated rod in a grounded metal case, and the glass windows at the front and back of the case allow for easy observation or projection. It is a good alternative for high humidity environments.
Specifications
  • Description:
    Gold foil electroscope