Gloeocapsa Slide
Gloeocapsa Slide, Whole Mount, Safranin
Catalog # 470177-416
Supplier: Avantor
Jump to:
Product Details & Documents
Cyanobacteria sheaths.
- Identifying features clearly distinguishable
- Rigorous quality control standards
Individuals and colonies in gelatinous capsules.
Specifications
- Description:Gloeocapsa Slide
- Stain:Safranin
- Orientation:Whole Mount