Order Entry
Mexico
ContactUsLinkComponent
Giardia lamblia Cysts Slide
Giardia lamblia Cysts Slide
Catalog # 470181-884
Supplier:  Avantor
undefined
Giardia lamblia Cysts Slide
Catalog # 470181-884
Supplier:  Avantor
Specifications
  • Description:
    Giardia lamblia Cysts
  • Cat. no.:
    470181-884
  • Stain:
    Hematoxylin
  • Supplier No.:
    WARD470181-884
  • Orientation:
    Whole mount
Specifications
About this item
Infectious protozoan species
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Common flagellated protozoan parasite. Causes giardiasis. Colonizes and reproduces in the small intestine. Most human infections occur from drinking sewage contaminated water.