Order Entry
Mexico
ContactUsLinkComponent
Generalized Plant Cell Slide
Catalog # 470177-554
Supplier:  Avantor
undefined
Generalized Plant Cell Slide
Catalog # 470177-554
Supplier:  Avantor
Specifications
  • Description:
    Generalized Plant Cell Slide
  • Cat. no.:
    470177-554
  • Stain:
    Quadruple
  • Supplier No.:
    WARD470177-554
  • Orientation:
    Section
Specifications
About this item
Investigate plant cell traits
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Cell wall, nucleus, nucleolus, large vacuole, and chloroplasts are shown. Study guide included.