General Examination of Urinary Sediment and Crystals Set
Catalog # 470183-652
Supplier: Avantor
Specifications
- Type:Animal
- Description:General Examination of Urinary Sediment and Crystals Set
- No. of Slides:8
- Cat. no.:470183-652
- Supplier No.:WARD470183-652
Specifications
About this item
WARD’S Exclusive. Demonstrates the Crystalline Forms Found in Urine
- Identifying features clearly distinguishable
- Rigorous quality control standards
All slides are whole mounts and includes stained and unstained urine sediment slides.
Ordering information: Set includes Sediments: unstained; SEDIstained; Crystals: Calcium oxalate; Calcium phosphate; Hippuric acid; Cystine; Uric acid; Triphosphate. The set also comes with the booklet General Examination of Urinary Sediments.