Order Entry
Mexico
ContactUsLinkComponent
Ward's® Gel Electrophoresis Simulation Lab Activity
Ward's® Gel Electrophoresis Simulation Lab Activity
Catalog # 470015-244
Proveedor:  Avantor
undefined
Ward's® Gel Electrophoresis Simulation Lab Activity
Catalog # 470015-244
Proveedor:  Avantor
Especificaciones
  • Description:
    Gel Electrophoresis Simulation Kit
  • No. of Lab Groups:
    15
  • Cat. no.:
    470015-244
  • Supplier No.:
    WARD470015-244
Especificaciones
Acerca de este artículo
Introduce Concept of Electrophoresis Without Hardware
  • "Dry Lab" to illustrate gel electrophoresis
  • Three activities included
  • Materials for 15 lab groups
  • Time required: ~60 minutes
Perfect for communicating the practice of electrophoresis simply and effectively without the need for electrophoresis equipment, this kit includes pop beads that students use to simulate the process of electrophoresis. Three investigations are included: assembling pop bead chains of amino acids to understand amino acids and protein structure, analyzing electrophoretic “results”, and determining DNA and protein fragment length. The kit includes enough materials for 15 setups, as well as a teacher’s guide and student copymaster.