Order Entry
Mexico
Orders LinkContactUsLinkComponent
Fungi Types Microscope Slide Set
Fungi Types Microscope Slide Set
Fungi Types Microscope Slide Set, 10, Fungi
Catalog # 470176-246
Supplier:   Avantor
Fungi Types Microscope Slide Set
Catalog # 470176-246
Supplier:  Avantor
Fungi Types Microscope Slide Set
Fungi Types Microscope Slide Set
Fungi Types Microscope Slide Set, 10, Fungi
Price
$2,109.60
Log in for availability
Kit de 1
Product Details & Documents
Study Both Sexual and Asexual Morphology in Fungi and Molds
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Especially useful to illustrate life cycles in different phylogenetic divisions within fungi, this slide set can also be used to compare and contrast with life cycles in other kingdoms.
Ordering information: Set includes: Lichen-mychobiont (wm); Ustilago maydis spore formation; Puccinia graminis - Uredia (sect); Puccinia graminis - Aecia (sect); Mushroom (cs); Budding yeast (wm); Blue mold - Penicillium (wm); Aspergillus (wm); Rhizopus nigricans with zygotes (wm); Physarum polycephalum plasmodium (cs).
Specifications
  • Type:
    Fungi
  • Description:
    Fungi Types Microscope Slide Set
  • No. of Slides:
    10