Fungi Survey Slide Set
Catalog # 470183-476
Supplier: Avantor
Specifications
- Type:Fungi
- Description:Fungi Survey Slide Set
- No. of Slides:10
- Cat. no.:470183-476
- Supplier No.:WARD470183-476
Specifications
About this item
Illustrates Examples From Each of the Major Phyla
- Identifying features clearly distinguishable
- Rigorous quality control standards
Ordering information: Set includes Zygomycetes: Rhizopus, sporangia and zygotes (wm); Ascomycetes: Yeast, three types (sm); Peziza, apothecia (sect); Basidomycetes: Mushroom Anatomy, coprinus (cs); Spore formation (sect); Polyporus pores, basidiocarp (sect); Lycoperdon, basidiocarp (ls); Deuteromycetes: Aspergillus, conidiophores (wm); Penicillium, conidiophore with conidia (wm); and Lichens: Foliose lichen (sect).