Order Entry
Mexico
ContactUsLinkComponent
Ward's® From DNA to a Protein Manipulatives
Ward's® From DNA to a Protein Manipulatives
  470105-974
 :  Avantor
undefined
Ward's® From DNA to a Protein Manipulatives
  470105-974
 :  Avantor
  • Description:
    From DNA to a Protein Manipulatives
  • No. of Lab Groups:
    10
  • Cat. no.:
    470105-974
  • Supplier No.:
    WARD470105-974
Hands on Protein synthesis
  • Pre and post-laboratory assessments
  • Links to real world concepts
  • Estimated class/lab time required: 30-45 minutes
Lead students through the process of protein synthesis using the ten sets of detailed laminated manipulatives in the kit, developed by Patti Richardson. Working in small groups or individually, students begin by bonding nucleotides to form a segment of DNA. They then use manipulatives representing mRNA, tRNA, a ribosome, bases, amino acids, and peptide bonds to work through the processes of transcription and translation, to synthesize a protein. Students also discover what happens if there is a mutation in the DNA and how this can alter the protein that is produced. Assembly required. Includes set of 10 and instructions. Grades 9–12.