Order Entry
Mexico
ContactUsLinkComponent
CENCO® Large Forced Oscillation and Resonance Demonstrator
CENCO® Large Forced Oscillation and Resonance Demonstrator
Catalog # 470121-166
undefined
CENCO® Large Forced Oscillation and Resonance Demonstrator
Catalog # 470121-166
Supplier Number:  SP761
Specifications
  • Description:
    Forced Oscillation & Resonance Demonstrator
  • Cat. no.:
    470121-166
  • Supplier No.:
    SP761
Specifications
About this item
Study Static and Dynamic Oscillations
Study Static and Dynamic Oscillations
This oversized apparatus is suitable for demonstrations in large classrooms, so all students can observe. Apply weights and note the spring elongations along the graduated scale. An acrylic glass cylinder, filled with oil or water, is used for study damping. The friction ratio can also be varied through changing discs. Excitation frequency is displayed on the LCD panel. Requires low voltage power supply available separately. Size: 35cm L x 35cm W x 85cm H.