Lab-Aids® Food Nutrient Analysis Kit
Catalog # 470111-220
Supplier: LAB-AIDS INC (KITS) SE
Specifications
- Description:Food Nutrient Analysis Kit
- Environmentally Preferable:
- No. of Lab Groups:30
- Cat. no.:470111-220
- Supplier No.:6A
Specifications
About this item
This kit identifies five classes of nutrients.
- Activities: Testing common foods and discussing conclusions
- Students quantitatively determine nutrients in foods
- Estimated lab time: one 50-minutes class period
- Team-centered activity
In this kit, students perform and standardize tests with pure samples of nutrients. Working in teams, students identify and test for five classes of nutrients: sugar, starch, lipid, protein, and Vitamin C.
Once students complete their tests and make observations, they discuss their conclusions on the results, controls, and samples that showed no reaction.
Ordering information: This kit includes a teacher's guide with SDS, student worksheets and guides, glucose, vitamin C, protein, starch suspension, vegetable oil, fat-testing solution, protein testing solution a, protein testing solution b, sugar-testing solution, vitamin c-testing solution, starch-testing solution, and test tubes. A hot-water bath, a large sheet of cardboard or dry-erase board, and a white screen are needed but not included.
Delivery: See datasheet for contents information.