Folding Pocket Magnifiers
Pocket Field Magnifier, Single Lens
Catalog # 470104-364
Supplier: UNITED SCIENTIFIC SUPPLIES
Supplier #: MPS038
Jump to:
Product Details & Documents
Sturdy plastic frames with 38mm (1.5") diameter lenses.
- Sturdy frame
- Folds up for storage
These versatile magnifiers can magnify objects up to 5x.
Specifications
- Magnification:5×
- Description:Pocket Field Magnifier, Single Lens
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



