Order Entry
Mexico
Orders LinkContactUsLinkComponent
Foam Brushes
Foam Brushes
Supplier:   BRUSHTECH
Foam Brushes
Supplier:  BRUSHTECH
Certificates
Foam Brushes
3 Options Available Below
Product Details & Documents
Foam brushes are designed for cleaning lab glass and plasticware.
  • Brushes will not chip, scratch or mar glassware
  • Handles are plastic, including grips
Brushes are made of a special foam that does not get wet, yet transports water and detergent to the glass surface. These brushes will easily out-live others. They should be a fixture in your prep room.
Product Family Options
Product Information
  • Bristle Material
    Handle Material
    Availability
    Price
  • 470004-700
    Foam Flask Brush
    Foam Flask Brush
    Foam
    Plastic
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $223.06
    $223.06
    Log in for availability
    Case de 48 (1UN)
    Case de 48 (1UN) - $243.13
    $243.13
    Each (1UN)
       $223.06
    Case de 48 (1UN)
       $243.13
    Supplier Number: B55C
  • 470004-702
    Foam Beaker Brush
    Foam Beaker Brush
    Foam
    Plastic
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $194.54
    $194.54
    Each (1UN)
       $194.54
    Supplier Number: B60C
  • 470120-528
    Foam Cylinder Brush
    Foam Cylinder Brush
    Foam
    Plastic
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $252.60
    $252.60
    Log in for availability
    Case de 12 (1UN)
    Case de 12 (1UN) - $275.33
    $275.33
    Each (1UN)
       $252.60
    Case de 12 (1UN)
       $275.33
    Supplier Number: B130C