Order Entry
Mexico
Orders LinkContactUsLinkComponent
Flat Plastic Meter Stick
Flat Plastic Meter Stick
Flat metric meter stick, Plastic, 1 m
  470047-182
Supplier #:  11050-A
Flat Plastic Meter Stick
  470047-182
Flat Plastic Meter Stick
Flat Plastic Meter Stick
Flat metric meter stick, Plastic, 1 m
$166.81
Each (1UN)
Flexible and flat, this durable plastic meter stick is marked in cm with 1 mm divisions on one side and with alternate-colored dm bars on the other, to illustrate the relationships between units.
Flexible and flat, this durable plastic meter stick is marked in cm with 1mm divisions on one side and with alternate-colored dm bars on the other, to illustrate the relationships between units.
  • Description:
    Flat metric meter stick
  • Length:
    1 m
  • Material:
    Plastic