Flask Form Electroscope
Catalog # 470193-152
Supplier: Avantor
Specifications
- Description:Flask Form Electroscope
- Cat. no.:470193-152
- Supplier No.:3132-01
Specifications
About this item
This inexpensive electroscope is housed in a 250 ml Pyrex flask, with aluminum leaves and a metal top knob.