Order Entry
Mexico
ContactUsLinkComponent
Ward's® Flame Test and Spectral Analysis Lab Activity
Ward's® Flame Test and Spectral Analysis Lab Activity
Catalog # 470012-260
Supplier:  Avantor
undefined
Ward's® Flame Test and Spectral Analysis Lab Activity
Catalog # 470012-260
Supplier:  Avantor
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    WARD’S Flame Test and Spectral Analysis Lab Activity
  • Cat. no.:
    470012-260
  • Supplier No.:
    WARD470012-260
Specifications
About this item
An Effective Introduction to Spectral Analysis
Astronomers learn important information about the composition of stars by analyzing their spectra. To gain an understanding of this process, students use a spectroscope to observe and describe the spectra of four different chemical salts when heated by a flame. The kit comes with four salts, four nichrome wire test loops, six spectroscopes, a teacher’s guide, and student copymasters. It includes enough materials for 30 or more tests.