Order Entry
Mexico
ContactUsLinkComponent
Filter Support Collar
Filter Support Collar
Catalog # 470017-722
undefined
Filter Support Collar
Catalog # 470017-722
Supplier Number:  FLTR01
Specifications
  • Description:
    Filter Support Collar
  • Cat. no.:
    470017-722
  • Supplier No.:
    FLTR01
Specifications
About this item
Filter Support Collar is a sturdy, molded rubber filter disc.
  • Inside Diameter: 32 mm
  • Outside Diameter: 76 mm
  • Molded Rubber Construction
It is designed to support and seal a filter crucible or funnel in the neck of a filter flask.