Fieldmaster Metal Sieve Set
470018-758
- Description:Fieldmaster Metal Sieve Set/6
- Cat. no.:470018-758
- Supplier No.:78-700
Sort and separate materials with this durable metal sieve set.
- Includes six sieves, pan, and cover Stacking set for simple storage Ideal for soil and material studies Strong, long-lasting material
Classify Materials with Precision Using the Fieldmaster Metal Sieve Set! Designed for geology, soil science, and material analysis, this durable set allows students and researchers to separate particles by size with ease. The stackable stainless steel mesh sieves come in six sizes—4000, 2000, 500, 250, 125, and 63 microns—making it simple to sort a wide range of materials. A lid and catch pan are included for clean, efficient collection and analysis. Rugged and economical, this set is a reliable tool for any lab or field study focused on particle classification.