Order Entry
Mexico
Orders LinkContactUsLinkComponent
Fever Tester
Fever Tester
Fever Tester
Catalog # 470146-260
Supplier #:  BFT45
Fever Tester
Catalog # 470146-260
Fever Tester
Fever Tester
Fever Tester
Price
$63.92
Log in for availability
Each (1UN) Case de 144 (1UN)
Product Details & Documents
Accurate Readings in Fifteen Seconds
Simply press this liquid crystal thermometer against the forehead and in 15 seconds the correct temperature is highlighted in green. No sterilization is required. Range: 95–104°F. Length: 3 1/2".
Specifications
  • Description:
    Fever Tester