Order Entry
Mexico
ContactUsLinkComponent
Families of Elements Experiment Kit
Families of Elements Experiment Kit
  470006-990
undefined
Families of Elements Experiment Kit
  470006-990
 :  110R-RC
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Families of Elements Experiment Kit, Refill
  • Cat. no.:
    470006-990
  • Supplier No.:
    110R-RC
Students Learn How to Classify Elements
  • Learn to classify elements by observing behavior of their compounds
  • Examine properties of the halogen compounds
  • Determine formulas of compounds through experimentation
  • Requires one 50 minute class period to complete
  • Number of lab groups: 15
Students learn how to classify elements by observing the behavior of compounds in the Halogen family (Group VIIA). The chemical properties of this group are very similar despite the marked differences in their physical properties. As students perform a series of experiments, they discover that the elements form remarkably similar compounds with silver. Also, that the formulas of the compounds are similar, and that the elements are indeed in the same family.
 : Kit includes Chemplates® with spatulas, LAB-AIDS®; Silver nitrate; Potassium bromide; Potassium chloride; Potassium iodide; Potassium sulfide.