Exploring With a Microscope II Slide Set
Exploring With a Microscope II Slide Set, 12
Catalog # 470183-640
Supplier: Avantor
Jump to:
Product Details & Documents
Includes an Illustrated Study Guide Highlighting Microscopic Features of “Common Things”
- All identifying features are clearly distinguishable
- Rigorous quality control standards
- In-house experts available to answer your questions
View familiar objects and materials at the microscopic level. All slides are whole mounts. This set comes with an illustrated study guide.
: Set includes Quartz sand; Cosmetic types; Dandruff; Cornstarch; Salt crystals; Nylon and Wool Fibers; Toothpaste; Human Hair; Sawdust; Iron rust, Common dust and Chalk.
Specifications
- Description:Exploring With a Microscope II Slide Set
- No. of Slides:12