Euglena Explano-Mount™ Slide
Euglena Explano-Mount™, Explano-Mount™
Catalog # 470183-644
Supplier: Avantor
Jump to:
Product Details & Documents
Slide and Illustrated Guide Packed in a Durable Plastic Sleeve
- Identifying features clearly distinguishable
- Rigorous quality control standards
General study. Demonstrates nucleus, chloroplasts, contractile vacuole, and flagellum.
Specifications
- Description:Euglena Explano-Mount™
- Orientation:Explano-Mount™