Order Entry
Mexico
Orders LinkContactUsLinkComponent
Etherizer
Etherizer
Etherizer, Drosophila, w/ Dropper Pipet
Catalog # 470154-018
Supplier:   Avantor
Etherizer
Catalog # 470154-018
Supplier:  Avantor
Etherizer
Etherizer, Drosophila, w/ Dropper Pipet
Price
$611.10
Log in for availability
Each (1UN)
Product Details & Documents
Specially designed unit to safely and easily etherize Drosophila or other living materials.
  • Etherized Drosophila drop into a collecting bottle
The bottle containing live material is inverted over the funnel. A few drops of ether are placed on a dispersing wick under the rubber plug.
Delivery information: Includes dropper pipet.
Specifications
  • Description:
    Etherizer, Drosophila, w/ Dropper Pipet