Order Entry
Mexico
Orders LinkContactUsLinkComponent
<i>Elodea</i>, Submerged Leaf Slide
Elodea, Submerged Leaf Slide
Elodea, Submerged Leaf, whole mount, Whole mount, Fuchsin and Fast Green
Catalog # 470177-458
Supplier:   Avantor
<i>Elodea</i>, Submerged Leaf Slide
Catalog # 470177-458
Supplier:  Avantor
<i>Elodea</i>, Submerged Leaf Slide
Elodea, Submerged Leaf Slide
Elodea, Submerged Leaf, whole mount, Whole mount, Fuchsin and Fast Green
Price
$178.80
Log in for availability
Each (1UN)
Product Details & Documents
Whole mount, fuchsin & fast green stain
  • All identifying features are clearly distinguishable
  • Rigorous quality control standards
Elodea, (water weed). Hydrophytic leaf, showing chloroplasts, nuclei, and cell walls. Demonstrates the two-cell layer construction of aquatic plants.
Specifications
  • Description:
    Elodea, Submerged Leaf, whole mount
  • Stain:
    Fuchsin and Fast Green
  • Orientation:
    Whole mount