Elodea, Stem Tip Slide
Elodea, Stem Tip Slide, Longitudinal section, Quadruple
470176-884
: Avantor
Illustrates new growth development
- Identifying features clearly distinguishable
- Rigorous quality control standards
Waterweed. Shows apical meristem and developing leaves.
- Description:Elodea, Stem Tip Slide
- Stain:Quadruple
- Orientation:Longitudinal section