Electromagnet Kit
Electromagnet Kit, 7 oz.
470039-946
: EMKIT2
Reliable kit in a quality carrying case!
- Build bar and U shaped electromagnets
- The kit includes a 5 x 5 x 1 1/2" plastic case
You will receive two coils on plastic spools, a U shaped iron core, a straight iron core, two magnetic needles with stands, and a pair of wires. Students can use these materials to build bar and U shaped electromagnets with a 6 V battery or DC power supply, and determine the magnitude and polarity of electromagnetic forces. Includes instructions.
- Description:Electromagnet Kit
- Weight:7 oz.