Order Entry
Mexico
Orders LinkContactUsLinkComponent
Electric Generator
Electric Generator
Generator, Electric
Catalog # 470006-534
Supplier #:  15
Electric Generator
Catalog # 470006-534
Electric Generator
Electric Generator
Generator, Electric
Price
$3,996.49
Log in for availability
Each (1UN)
Product Details & Documents
This belt-driven generator may be used with our Steam Engine (63030-00 ). Operating at approximately 1500 rpm, the generator will produce enough electricity to light the included 3.8 volt bulb. The unit is mounted on a metal base (7.6×11.4 cm) and has a V-grooved pulley for the drive belt.
Specifications
  • Description:
    Generator, Electric