Order Entry
Mexico
ContactUsLinkComponent
Dual Calibration Spring Scales
Dual Calibration Spring Scales
Catalog # 470008-100
undefined
Dual Calibration Spring Scales
Catalog # 470008-100
Supplier Number:  1712601
Specifications
  • Description:
    Dual calibration spring scale, green
  • Environmentally Preferable:
  • Cat. no.:
    470008-100
  • Weighing Capacity:
    500 g/5 N
  • Supplier No.:
    1712601
Specifications
About this item
Economy Color-Coded Spring Scales
  • Provide direct readings for both force and mass measurements
  • Calibrated in both grams and Newtons
  • Can be fully zeroed
The body tube is constructed of clear plastic with color-coded plastic ends, and chrome-plated hooks and rings. The top of each spring is leveled for easy use on an inclined plane.
Caution: Caution:CHOKING HAZARD–These scales contain small parts and are not for children under three years.