Doublet Magnifier
Doublet Magnifier
Catalog # 470122-662
Supplier: UNITED SCIENTIFIC SUPPLIES
Supplier #: MPF010
Jump to:
Product Details & Documents
This compact, highly corrected magnifier comes in a convenient folding holder and case.
- Diameter: 1.3 cm
- Focal Length: 2.7 cm
- Magnification: 10x
- Convenient Folding Holder and Case
- Highly Corrected
It is 1.3 cm in diameter and has a focal length of 2.7 cm.
The magnification is 10x.
Specifications
- Magnification:10×
- Description:Doublet Magnifier
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



