Order Entry
Mexico
ContactUsLinkComponent
Dissectible Coils and Cores
Dissectible Coils and Cores
Catalog # 470120-754
Supplier:  VWR International
undefined
Dissectible Coils and Cores
Catalog # 470120-754
Supplier:  VWR International
Specifications
  • Description:
    Dissectible Coils and Cores
  • Cat. no.:
    470120-754
  • Supplier No.:
    PH0850CSET
Specifications
About this item
This set features five coils.
  • Coils Illustrate Electromagnetism and Induction
  • Useful in Basic Transformer Experiments
  • Included Coils Have 200, 400, 800, 1600, and 3200 Turns
  • Five Coils in Set
The coils illustrate electromagnetism and induction. They are also useful in basic transformer experiments.
The included coils have 200, 400, 800, 1600, and 3200 turns.