Order Entry
Mexico
Orders LinkContactUsLinkComponent
Digital Solar Power Meter

Digital Solar Power Meter

Digital Solar Power Meter, 4.3", 2.5", 1.3"
Catalog # 470018-066
Supplier #:  DBTU1300
Digital Solar Power Meter
Catalog # 470018-066
Digital Solar Power Meter
Digital Solar Power Meter

Digital Solar Power Meter, 4.3", 2.5", 1.3"

Price
$5,266.12
Log in for availability
Each (1UN)
Product Details & Documents

Ideal for checking thermal efficiency

  • Measures and displays solar power intensity in units of W/m2 or Btu/(ft2 x h)
  • Wide spectral range
  • Uses silicon photodiode and cosine angular correction
  • Min/max/avg memory + data hold
  • Stores 99 readings

This meter displays four digit readings on a large LCD screen. Its wide spectral range offers accurate solar energy measurements using a silicon photovoltaic detector. Select either W/m2 or Btu / (ft2 x h) units. Additional features include data hold, minimum/maximum/average values, and auto-off. Includes four AAA 1.5V batteries, tripod mounting screw, and carrying case.

Specifications: Range: 2000 W/m2 , 634 Btu / (ft2 x h). Resolution: 0.1 W/m2 , 0.1 Btu / (ft2 x h). Spectral Response: 400 to 1000nm. Angular Accuracy: Cosine corrected <5% for angles <60°. Operating Temperature and Humidity: 0 to 50°C; 32 to 122°F; 0% to 80% RH. Size: 4.3" x 2.5" x 1.3". Weight: 5.6 oz.

Specifications
  • Description:
    Digital Solar Power Meter
  • Height:
    1.3"
  • Length:
    4.3"
  • Width:
    2.5"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.